Learn More
Medchemexpress LLC 4-Methoxyphenethyl alcohol | 702-23-8 | 152.19 | 100 G
SDP
Catalog No. 5000445673
Encompass_Preferred
4-Methoxyphenethyl alcohol is an aromatic alcohol that is the primary component contributing to the anise-like odor produced by Amorphophallus. This compound has been shown to inhibit protein, RNA, and DNA synthesis in Escherichia coli. It exhibits a complete bacteriostatic concentration of 4.8 mM against Escherichia coli, concurrently inhibiting the synthesis of DNA, RNA, and protein.
  • Primary component for anise-like odor in Amorphophallus
  • Inhibits protein, RNA, and DNA synthesis
  • Exhibits bacteriostatic concentration against Escherichia coli
  • Concurrent inhibition of DNA, RNA, and protein synthesis
Catalog No. 50-004-45673 Supplier Medchemexpress LLC Supplier No. HYW004056100G
4-Methoxyphenethyl alcohol is an aromatic alcohol that is the primary component contributing to the anise-like odor produced by Amorphophallus. This compound has been shown to inhibit protein, RNA, and DNA synthesis in Escherichia coli. It exhibits a complete bacteriostatic concentration of 4.8 mM against Escherichia coli, concurrently inhibiting the synthesis of DNA, RNA, and protein.
  • Primary component for anise-like odor in Amorphophallus
  • Inhibits protein, RNA, and DNA synthesis
  • Exhibits bacteriostatic concentration against Escherichia coli
  • Concurrent inhibition of DNA, RNA, and protein synthesis