Learn More
Medchemexpress LLC 4-Methoxyphenethyl alcohol | 702-23-8 | 152.19 | 25 G
SDP
Catalog No. 5000448282
Encompass_Preferred
4-Methoxyphenethyl alcohol is an aromatic alcohol and the primary component of the anise-like odor produced by Amorphophallus. It can inhibit the synthesis of protein, RNA, and DNA in *Escherichia coli*. In vitro, it exhibits a complete bacteriostatic concentration of 4.8 mM against *Escherichia coli*, concurrently inhibiting the synthesis of DNA, RNA, and protein.
  • Aromatic alcohol
  • Primary component of anise-like odor from Amorphophallus
  • Inhibits synthesis of protein, RNA, and DNA in *Escherichia coli*
  • Exhibits bacteriostatic concentration of 4.8 mM against *Escherichia coli*
Catalog No. 50-004-48282 Supplier Medchemexpress LLC Supplier No. 25GHYW004056
4-Methoxyphenethyl alcohol is an aromatic alcohol and the primary component of the anise-like odor produced by Amorphophallus. It can inhibit the synthesis of protein, RNA, and DNA in *Escherichia coli*. In vitro, it exhibits a complete bacteriostatic concentration of 4.8 mM against *Escherichia coli*, concurrently inhibiting the synthesis of DNA, RNA, and protein.
  • Aromatic alcohol
  • Primary component of anise-like odor from Amorphophallus
  • Inhibits synthesis of protein, RNA, and DNA in *Escherichia coli*
  • Exhibits bacteriostatic concentration of 4.8 mM against *Escherichia coli*