Learn More
Medchemexpress LLC 4-Methoxyphenethyl alcohol | 702-23-8 | 152.19 | 50 G
SDP
Catalog No. 5000445856
Encompass_Preferred
4-Methoxyphenethyl alcohol is an aromatic alcohol, identified as a major component of the anise-like odor produced by Amorphophallus. It inhibits protein, RNA, and DNA synthesis, demonstrating a complete bacteriostatic concentration of 4.8 mM against Escherichia coli.
  • Aromatic alcohol
  • Major component of anise-like odor
  • Inhibits protein, RNA, and DNA synthesis
  • Bacteriostatic against Escherichia coli at 4.8 mM
Catalog No. 50-004-45856 Supplier Medchemexpress LLC Supplier No. HYW00405650G
4-Methoxyphenethyl alcohol is an aromatic alcohol, identified as a major component of the anise-like odor produced by Amorphophallus. It inhibits protein, RNA, and DNA synthesis, demonstrating a complete bacteriostatic concentration of 4.8 mM against Escherichia coli.
  • Aromatic alcohol
  • Major component of anise-like odor
  • Inhibits protein, RNA, and DNA synthesis
  • Bacteriostatic against Escherichia coli at 4.8 mM