Learn More
Medchemexpress LLC 4-Methoxyphenethyl alcohol | 702-23-8 | MFCD00002900 | 152.19 | 5 G
SDP
Catalog No. 5000447142
Encompass_Preferred
4-Methoxyphenethyl alcohol is an aromatic alcohol and a primary component of the anise-like odor produced by *Amorphophallus*. This compound has been shown to inhibit the synthesis of protein, RNA, and DNA in *Escherichia coli*.
  • Aromatic alcohol responsible for anise-like odor in *Amorphophallus*.
  • Inhibits protein, RNA, and DNA synthesis in *Escherichia coli*.
  • Exhibits a complete bacteriostatic concentration of 4.8 mM against *Escherichia coli*.
  • Available in solid or liquid form.
  • For research use only.
Catalog No. 50-004-47142 Supplier Medchemexpress LLC Supplier No. HYW0040565G
4-Methoxyphenethyl alcohol is an aromatic alcohol and a primary component of the anise-like odor produced by *Amorphophallus*. This compound has been shown to inhibit the synthesis of protein, RNA, and DNA in *Escherichia coli*.
  • Aromatic alcohol responsible for anise-like odor in *Amorphophallus*.
  • Inhibits protein, RNA, and DNA synthesis in *Escherichia coli*.
  • Exhibits a complete bacteriostatic concentration of 4.8 mM against *Escherichia coli*.
  • Available in solid or liquid form.
  • For research use only.