Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenethyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. M088925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M088925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M088925G Supplier TCI America Supplier No. M088925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 702-23-8
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00002900
InChI Key AUWDOZOUJWEPBA-UHFFFAOYSA-N
Synonym 2-4-methoxyphenyl ethanol, 4-methoxyphenethyl alcohol, 4-methoxybenzeneethanol, benzeneethanol, 4-methoxy, p-methoxyphenethyl alcohol, 2-4-methoxyphenyl ethan-1-ol, unii-3mwl5nze9k, 3mwl5nze9k, 4-2-hydroxyethyl anisole, 2-4-methoxyphenyl-ethanol
PubChem CID 69705
IUPAC Name 2-(4-methoxyphenyl)ethanol
SMILES COC1=CC=C(C=C1)CCO

Specifications

Color White-Yellow
Boiling Point 336°C
Quantity 25 g
Formula Weight 152.19
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 4-Methoxyphenethyl Alcohol
EINECSNumber (3)-4080
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.