Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Methoxyphenol

Catalog No. M186555G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
50 lb.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M186555G 5 g
M1865550LB 50 lb.
M18655100G 100 g
M186551KG 1 kg
M18655500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. M186555G Supplier Sigma Aldrich Supplier No. M186555G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Methoxyphenol

Specifications

CAS 150-76-5
Melting Point 55°C to 57°C (lit.)
Boiling Point 243°C (lit.)
Molecular Formula C7H8O2
Linear Formula CH3OC6H4OH
MDL Number MFCD00002332
Quantity 5 g
Synonym p-Methoxyphenol; 4-Hydroxyanisole; 4-MP; HQMME; Hydroquinone monomethyl ether; MEHQ; MQ-F; p-Guaiacol
Molecular Weight (g/mol) 124.14
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.