Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger TCI AMERICA 4-Methoxyphenol, 99.0% Purity, 25 g

Catalog No. 502781461
Encompass_Preferred

4-Methoxyphenol is a phenol-based building block commonly used in organic synthesis, particularly for the protection and deprotection of PMP groups. This analytical reagent is supplied as a white to light yellow or light orange powder or crystal.

  • Chemical formula: C7H8O2
  • Molecular weight: 124.14
  • Purity: ≥99.0% (GC)
  • Melting point: 55.0°C to 58.0°C
  • Boiling point: 243°C
  • Solubility: Soluble in water (40,000 mg/L at 25°C), benzene, methanol, and ethanol
  • Physical state: Solid
  • Storage: Room temperature (recommended <15°C) under inert gas
  • Sensitivity: Air sensitive

Catalog No. 50-278-1461 Supplier Grainger Supplier No. 19KT67
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Methoxyphenol is a phenol-based building block commonly used in organic synthesis, particularly for the protection and deprotection of PMP groups. This analytical reagent is supplied as a white to light yellow or light orange powder or crystal.

  • Chemical formula: C7H8O2
  • Molecular weight: 124.14
  • Purity: ≥99.0% (GC)
  • Melting point: 55.0°C to 58.0°C
  • Boiling point: 243°C
  • Solubility: Soluble in water (40,000 mg/L at 25°C), benzene, methanol, and ethanol
  • Physical state: Solid
  • Storage: Room temperature (recommended <15°C) under inert gas
  • Sensitivity: Air sensitive

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.