Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger TCI 4-Methoxyphenol, >99.0% (GC), Solid, 500 g

Catalog No. 502781609
Encompass_Preferred

4-Methoxyphenol, also known as hydroquinone monomethyl ether, is a reagent-grade chemical utilized primarily for the protection and deprotection of PMP groups in laboratory research. This solid compound is characterized by high purity levels.

  • Purity: ≥99.0% (GC)
  • Molecular formula: C7H8O2
  • Molecular weight: 124.14 g/mol
  • Physical state: Solid
  • Melting point: 57°C
  • Boiling point: 243°C
  • Solubility: Soluble in water, benzene, methanol, and ethanol
  • Storage: Room temperature (recommended in a cool and dark place, <15°C); store under inert gas
  • Sensitive to air

Catalog No. 50-278-1609 Supplier Grainger Supplier No. 19KT68
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Methoxyphenol, also known as hydroquinone monomethyl ether, is a reagent-grade chemical utilized primarily for the protection and deprotection of PMP groups in laboratory research. This solid compound is characterized by high purity levels.

  • Purity: ≥99.0% (GC)
  • Molecular formula: C7H8O2
  • Molecular weight: 124.14 g/mol
  • Physical state: Solid
  • Melting point: 57°C
  • Boiling point: 243°C
  • Solubility: Soluble in water, benzene, methanol, and ethanol
  • Storage: Room temperature (recommended in a cool and dark place, <15°C); store under inert gas
  • Sensitive to air

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.