Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenol, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013195 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3195 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3195 Supplier MilliporeSigma Supelco Supplier No. 92264250MG
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 789°F
CAS 150-76-5
Vapor Pressure <0.01 mmHg (20°C)
Vapor Density 4.3 (vs. air)
Flash Point 132°C
Physical Form Neat
Linear Formula CH3 OC6H4 OH
MDL Number MFCD00002332
Melting Point 55°C to 57°C
Boiling Point 243°C
Synonym 4-Hydroxyanisole; 4-MP; HQMME; Hydroquinone monomethyl ether; MEHQ; MQ-F; p-Guaiacol
Type Analytical Standard
Quantity 250 mg
Formula Weight 124.14
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.