Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenol, 99%

Catalog No. AC126000250
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
25 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC126000250 25 kg
AC126001000 100 g
AC126005000 500 g
AC126000010 1 kg
AC126000051 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC126000250 Supplier Thermo Scientific Chemicals Supplier No. 126000250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 150-76-5 | C7H8O2 | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-76-5 , 123-31-9
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002332
InChI Key NWVVVBRKAWDGAB-UHFFFAOYSA-N
Synonym mequinol, 4-hydroxyanisole, p-hydroxyanisole, p-methoxyphenol, hydroquinone monomethyl ether, phenol, 4-methoxy, leucobasal, mehq, leucodine b, mechinolum
PubChem CID 9015
ChEBI CHEBI:69441
IUPAC Name 4-methoxyphenol
SMILES COC1=CC=C(O)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 56.0°C to 57.0°C
Color Colorless to Yellow
Boiling Point 243.0°C
Flash Point 133°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Quantity 25 kg
Beilstein 06,843
Solubility Information Solubility in water: 40g/L (25°C). Other solubilities: readily soluble in ethyl acetate, 455g/100g ethyl alcohol - 426g/100g acetone (25°C), 70g/100g benzene - 3g/100g carbon tetrachloride, (25°C)
Viscosity 8.5 mPa.s (65°C)
Formula Weight 124.14
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material 4-Methoxyphenol, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of damaging the unborn child.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
If eye i
EINECSNumber 205-769-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.