Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenyl 2,3,4,6-Tetra-O-acetyl-beta-D-galactopyranoside 98.0+%, TCI America™
SDP

Catalog No. M14775G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M14775G Supplier TCI America Supplier No. M14775G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2872-65-3 | C21H26O11 | 454.428 g/mol

Chemical Identifiers

CAS 2872-65-3
Molecular Formula C21H26O11
Molecular Weight (g/mol) 454.428
MDL Number MFCD06797123
InChI Key RPHXBVOPPUTUES-XDWAVFMPSA-N
PubChem CID 13325756
IUPAC Name [(2R,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-(4-methoxyphenoxy)oxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC2=CC=C(C=C2)OC)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 5 g
Formula Weight 454.43
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 2,3,4,6-Tetra-O-acetyl-beta-D-galactopyranoside
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.