Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenyl 2-Azido-3,6-di-O-benzyl-2-deoxy-beta-D-glucopyranoside, TCI America™
SDP

Catalog No. M16171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M16171G Supplier TCI America Supplier No. M16171G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A sugar azide used in conjugation and click chemistry research.

Chemical Identifiers

CAS 1272755-25-5
Molecular Formula C27H29N3O6
Molecular Weight (g/mol) 491.544
InChI Key ZTAXYCGZXSYIMV-RFNQJFSXSA-N
PubChem CID 53384632
IUPAC Name (2R,3S,4R,5R,6S)-5-azido-6-(4-methoxyphenoxy)-4-phenylmethoxy-2-(phenylmethoxymethyl)oxan-3-ol
SMILES COC1=CC=C(C=C1)OC2C(C(C(C(O2)COCC3=CC=CC=C3)O)OCC4=CC=CC=C4)N=[N+]=[N-]

Specifications

Color White
Quantity 1 g
Formula Weight 491.54
Percent Purity >98.0%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 2-Azido-3,6-di-O-benzyl-2-deoxy-beta-D-glucopyranoside
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.