Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenyl 3-O-Benzyl-4,6-O-benzylidene-2-deoxy-2-phthalimido-beta-D-glucopyranoside 98.0+%, TCI America™
SDP

Catalog No. M16091G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M16091G Supplier TCI America Supplier No. M16091G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 129575-88-8
Molecular Formula C35H31NO8
Molecular Weight (g/mol) 593.632
MDL Number MFCD06797139
InChI Key WCCGRDKROUUDBC-LPRGBYEPSA-N
PubChem CID 126969985
IUPAC Name 2-[(4aR,6R,7R,8R,8aS)-6-(4-methoxyphenoxy)-2-phenyl-8-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-7-yl]isoindole-1,3-dione
SMILES COC1=CC=C(C=C1)OC2C(C(C3C(O2)COC(O3)C4=CC=CC=C4)OCC5=CC=CC=C5)N6C(=O)C7=CC=CC=C7C6=O

Specifications

Melting Point 133°C
Color White-Yellow
Quantity 1 g
Formula Weight 593.63
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 3-O-Benzyl-4,6-O-benzylidene-2-deoxy-2-phthalimido-beta-D-glucopyranoside
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.