Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenyl 4,6-O-Benzylidene-2-deoxy-2-phthalimido-beta-D-glucopyranoside 98.0+%, TCI America™
SDP

Catalog No. M14795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M14795G Supplier TCI America Supplier No. M14795G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 138906-43-1
Molecular Formula C28H25NO8
Molecular Weight (g/mol) 503.51
MDL Number MFCD06797124
InChI Key SHSWKRFHEJOKLK-UHFFFAOYNA-N
PubChem CID 126969997
IUPAC Name 2-[8-hydroxy-6-(4-methoxyphenoxy)-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-7-yl]-2,3-dihydro-1H-isoindole-1,3-dione
SMILES COC1=CC=C(OC2OC3COC(OC3C(O)C2N2C(=O)C3=CC=CC=C3C2=O)C2=CC=CC=C2)C=C1

Specifications

Melting Point 142°C
Color White
Quantity 5 g
Formula Weight 503.51
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 4,6-O-Benzylidene-2-deoxy-2-phthalimido-beta-D-glucopyranoside
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.