Learn More
AdipoGen 4-Methoxyphenyl isocyanide
SDP
Catalog No. 502560446
Encompass_Preferred
Chemical. CAS 10349-38-9. Formula C8H7NO. MW BD9837. 4-Methoxyphenyl isocyanide is a buildig block and intermediate for organic synthesis. This compound is colorless and volatile liquid when maintained at room temperature and it is extensively used in synthesis for many different bioreagents.
Catalog No. 50-256-0446 Supplier AdipoGen Supplier No. CDXM0730G005
Chemical. CAS 10349-38-9. Formula C8H7NO. MW BD9837. 4-Methoxyphenyl isocyanide is a buildig block and intermediate for organic synthesis. This compound is colorless and volatile liquid when maintained at room temperature and it is extensively used in synthesis for many different bioreagents.