Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-methoxyphenyl isothiocyanate | 2284-20-0 | MFCD00011676 | 165.21 g/mol | C8H7NOS | 1 G
SDP

Catalog No. 5000284471
Encompass_Preferred

4-Methoxyphenyl isothiocyanate is an organic isothiocyanate reagent used in biochemical research to evaluate antioxidant properties and cholinesterase interactions. It exhibits measurable radical-scavenging activity and partial inhibition of cholinesterase enzymes, and is supplied in small laboratory pack sizes for research applications.

  • CAS number 2284-20-0.
  • Molecular formula C8H7NOS; molecular weight 165.21 g/mol.
  • Reported antioxidant activity: DPPH IC50 = 1.25 mM; ORAC = 11.7 mM Trolox equivalents.
  • Partial cholinesterase inhibition: acetylcholinesterase ≈30.4%; butyrylcholinesterase ≈17.9%.
  • Available pack sizes: 1 g, 5 g, 10 g, 50 g.
  • Appearance: colorless to light yellow; solid-liquid mixture.

Catalog No. 50-002-84471 Supplier Medchemexpress LLC Supplier No. HYW5402321G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

4-Methoxyphenyl isothiocyanate is an organic isothiocyanate reagent used in biochemical research to evaluate antioxidant properties and cholinesterase interactions. It exhibits measurable radical-scavenging activity and partial inhibition of cholinesterase enzymes, and is supplied in small laboratory pack sizes for research applications.

  • CAS number 2284-20-0.
  • Molecular formula C8H7NOS; molecular weight 165.21 g/mol.
  • Reported antioxidant activity: DPPH IC50 = 1.25 mM; ORAC = 11.7 mM Trolox equivalents.
  • Partial cholinesterase inhibition: acetylcholinesterase ≈30.4%; butyrylcholinesterase ≈17.9%.
  • Available pack sizes: 1 g, 5 g, 10 g, 50 g.
  • Appearance: colorless to light yellow; solid-liquid mixture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.