Learn More
Medchemexpress LLC 4-methoxyphenyl isothiocyanate | 2284-20-0 | MFCD00011676 | 165.21 g/mol | C8H7NOS | 5 G
SDP
Catalog No. 5000284979
Encompass_Preferred
4-Methoxyphenyl isothiocyanate is an organic research reagent used as an electrophilic isothiocyanate building block for synthesis and derivatization. It is supplied in small laboratory quantities and should be handled and stored according to the safety data sheet.
  • Electrophilic isothiocyanate for introducing thiourea and related motifs.
  • Useful building block for small-molecule synthesis and derivatization.
  • Applicable in medicinal chemistry and reagent screening workflows.
  • Small, easy-to-handle 5 g laboratory package.
  • Handle per SDS recommendations; use appropriate personal protective equipment.
Catalog No. 50-002-84979 Supplier Medchemexpress LLC Supplier No. HYW5402325G
4-Methoxyphenyl isothiocyanate is an organic research reagent used as an electrophilic isothiocyanate building block for synthesis and derivatization. It is supplied in small laboratory quantities and should be handled and stored according to the safety data sheet.
  • Electrophilic isothiocyanate for introducing thiourea and related motifs.
  • Useful building block for small-molecule synthesis and derivatization.
  • Applicable in medicinal chemistry and reagent screening workflows.
  • Small, easy-to-handle 5 g laboratory package.
  • Handle per SDS recommendations; use appropriate personal protective equipment.