Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenyl Isothiocyanate 98.0+%, TCI America™
SDP

Catalog No. I05135G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I05135G Supplier TCI America Supplier No. I05135G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 2284-20-0
Melting Point 18°C
Color Yellow
Boiling Point 281°C
Molecular Formula C8H7NOS
Quantity 5 g
MDL Number MFCD00011676
UN Number 1760
Synonym 4-methoxyphenyl isothiocyanate, 4-methoxyphenylisothiocyanate, 1-isothiocyanato-4-methoxy-benzene, p-methoxyphenyl isothiocyanate, benzene, 1-isothiocyanato-4-methoxy, isothiocyanic acid 4-methoxyphenyl ester, 4-methoxybenzenisothiocyanate, 4-isothiocyanatoanisole, acmc-1ccqx, p-methoxyphenylisothiocyanate
InChI Key VRPQCVLBOZOYCG-UHFFFAOYSA-N
SMILES COC1=CC=C(C=C1)N=C=S
IUPAC Name 1-isothiocyanato-4-methoxybenzene
Molecular Weight (g/mol) 165.21
PubChem CID 75293
Formula Weight 165.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methoxyphenyl Isothiocyanate
Show More Show Less
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.