Learn More
Medchemexpress LLC 4-Methoxyphenylboronic acid | 5720-07-0 | 151.96 | 25 G
SDP
Catalog No. 5000448046
Encompass_Preferred
4-Methoxyphenylboronic acid is a white to off-white solid, biochemical reagent for life science research. It serves as a biological material or organic compound, identified by CAS number 5720-07-0 and a molecular weight of 151.96. The product is available in various quantities, typically with high purity, making it suitable for a wide range of laboratory applications.
  • Suitable for life science research
  • Functions as a biological material or organic compound
  • Available as a white to off-white solid
  • High purity for reliable experimental results
  • Solid form stores at room temperature for 3 years
  • In-solvent storage at -80°C for 2 years or -20°C for 1 year
  • Avoids inactivation from repeated freeze-thaw cycles by aliquoting solutions
  • Soluble in DMSO (100 mg/mL)
Catalog No. 50-004-48046 Supplier Medchemexpress LLC Supplier No. 25GHYW002177
Add to Cart
Add to Cart
4-Methoxyphenylboronic acid is a white to off-white solid, biochemical reagent for life science research. It serves as a biological material or organic compound, identified by CAS number 5720-07-0 and a molecular weight of 151.96. The product is available in various quantities, typically with high purity, making it suitable for a wide range of laboratory applications.
  • Suitable for life science research
  • Functions as a biological material or organic compound
  • Available as a white to off-white solid
  • High purity for reliable experimental results
  • Solid form stores at room temperature for 3 years
  • In-solvent storage at -80°C for 2 years or -20°C for 1 year
  • Avoids inactivation from repeated freeze-thaw cycles by aliquoting solutions
  • Soluble in DMSO (100 mg/mL)