Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Methoxyphenylboronic acid | 5720-07-0 | 151.96 | 250 G
SDP

Catalog No. 5000449943
Encompass_Preferred

4-Methoxyphenylboronic acid is a biochemical reagent used in life science research as a biological material or organic compound. It is a solid, typically white to off-white in appearance, with a molecular weight of 151.96 and a CAS number of 5720-07-0. This product is intended for research use only.

  • Appears as a solid
  • Color is white to off-white
  • Ships at room temperature in continental US; may vary elsewhere
  • Store at room temperature for 3 years (solid)
  • Store in solvent at -80°C for 2 years or -20°C for 1 year
  • Soluble in DMSO at 100 mg/mL (658.07 mM)

Catalog No. 50-004-49943 Supplier Medchemexpress LLC Supplier No. 250GHYW002177
May include imposed supplier surcharges.
Add to Cart
Add to Cart

4-Methoxyphenylboronic acid is a biochemical reagent used in life science research as a biological material or organic compound. It is a solid, typically white to off-white in appearance, with a molecular weight of 151.96 and a CAS number of 5720-07-0. This product is intended for research use only.

  • Appears as a solid
  • Color is white to off-white
  • Ships at room temperature in continental US; may vary elsewhere
  • Store at room temperature for 3 years (solid)
  • Store in solvent at -80°C for 2 years or -20°C for 1 year
  • Soluble in DMSO at 100 mg/mL (658.07 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.