Learn More
Cayman Chemical 4-Methoxyphenyl-β-D-Glucopyranoside, Purity: ≥98%, CAS: 6032-32-2, 5 g
SDP
Catalog No. 502912715
Encompass_Preferred
An aryl glycoside with ice recrystallization inhibitory activity; reduces mean grain size in a cell-free ice recrystallization assay and increases the percentage of intact isolated RBCs in PBS post-freeze-thaw at 100 mM; an intermediate in the synthesis of glucan oligosaccharides
Catalog No. 50-291-2715 Supplier Cayman Chemical Supplier No. 358185 G
An aryl glycoside with ice recrystallization inhibitory activity; reduces mean grain size in a cell-free ice recrystallization assay and increases the percentage of intact isolated RBCs in PBS post-freeze-thaw at 100 mM; an intermediate in the synthesis of glucan oligosaccharides