Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Methoxyphenylmagnesium Bromide Solution

Catalog No. 4702601L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-026-01L 1 L
47-026-0100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 47-026-01L Supplier Sigma Aldrich Supplier No. 4702601L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Methoxyphenylmagnesium bromide is a general Grignard reagent that can be used:In the denickelation of porphyrins and to replace the metal center with magnesium.To synthesize a tubulin polymerization inhibitor, 2-Amino-3,4,5-trimethoxybenzophenone.In the total synthesis of (-)-centrolobine.

Specifications

CAS 13139-86-1
Density 0.955 g/mL at 25°C
Boiling Point 65°C to 67°C
Molecular Formula C7H7BrMgO
Linear Formula CH3OC6H4MgBr
MDL Number MFCD00013515
Quantity 1 L
Molecular Weight (g/mol) 211.34 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.