Learn More
Research Products International Corp 4-Methylumbelliferyl-N-acetyl-B-D-glucosaminide, 100 MG
SDP
Catalog No. 501978195
Encompass_Preferred
Fluorescent substrate for N-Acetyl-B-D-glucosaminidase and B-N-Acetyl-D-hexosaminidase. Ultrasensitive. Checked by actual enzymatic assay.
Catalog No. 50-197-8195 Supplier Research Products International Corp Supplier No. M641000.1
Fluorescent substrate for N-Acetyl-B-D-glucosaminidase and B-N-Acetyl-D-hexosaminidase. Ultrasensitive. Checked by actual enzymatic assay.