Learn More
Medchemexpress LLC 4-Methyl-1,3-benzodioxole-5-carboxylic acid | 162506-58-3 | 96.28% | 180.16 | 10 G
SDP
Catalog No. 5000404689
Encompass_Preferred
4-Methyl-1,3-benzodioxole-5-carboxylic acid is a drug intermediate for the synthesis of various active compounds. It is intended for research use only.
  • Solid appearance
  • White to off-white color
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-04689 Supplier Medchemexpress LLC Supplier No. HY4140510G
4-Methyl-1,3-benzodioxole-5-carboxylic acid is a drug intermediate for the synthesis of various active compounds. It is intended for research use only.
  • Solid appearance
  • White to off-white color
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month