Learn More
Medchemexpress LLC 4-Methyl-1,3-benzodioxole-5-carboxylic acid | 162506-58-3 | 98.04% | 180.16 | 250 MG
SDP
Catalog No. 5000402313
Encompass_Preferred
4-Methyl-1,3-benzodioxole-5-carboxylic acid is a drug intermediate for the synthesis of various active compounds. It is for research use only and not sold to patients.
  • Drug intermediate for synthesis of various active compounds.
  • Available in different quantities including 250 mg.
  • Solid appearance, white to off-white color.
  • Storage recommendations: Powder -20°C for 3 years, 4°C for 2 years; In solvent -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-004-02313 Supplier Medchemexpress LLC Supplier No. 250MGHY41405
4-Methyl-1,3-benzodioxole-5-carboxylic acid is a drug intermediate for the synthesis of various active compounds. It is for research use only and not sold to patients.
  • Drug intermediate for synthesis of various active compounds.
  • Available in different quantities including 250 mg.
  • Solid appearance, white to off-white color.
  • Storage recommendations: Powder -20°C for 3 years, 4°C for 2 years; In solvent -80°C for 6 months, -20°C for 1 month.