Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-1-cyclohexen-1-ylboronic acid, 97%

Catalog No. AC431430010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC431430010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC431430010 Supplier Thermo Scientific Chemicals Supplier No. 431430010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 850567-92-9 | C7H13BO2 | 139.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850567-92-9
Molecular Formula C7H13BO2
Molecular Weight (g/mol) 139.99
MDL Number MFCD06659857
InChI Key SJZMIZIVYIOMIW-UHFFFAOYNA-N
Synonym 4-methyl-1-cyclohexen-1-ylboronic acid, 4-methylcyclohex-1-en-1-yl boronic acid, 4-methylcyclohex-1-en-1-ylboronic acid, 4-methylcyclohex-1-enylboronic acid, boronic acid, 4-methyl-1-cyclohexen-1-yl, boronic acid,b-4-methyl-1-cyclohexen-1-yl, acmc-209q0u, 4-methyl cyclohexen-1-ylboronic acid, 4-methylcyclohexen-1-yl boronic acid, 4-methyl-1-cyclohexenyl boronic acid
PubChem CID 44119556
IUPAC Name (4-methylcyclohexen-1-yl)boronic acid
SMILES CC1CCC(=CC1)B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 155.0°C to 159.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 139.99
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-1-cyclohexen-1-ylboronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.