Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-1-cyclohexene, TCI America™
SDP

Catalog No. M22015G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M22015G Supplier TCI America Supplier No. M22015G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 591-47-9
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00001573
InChI Key FSWCCQWDVGZMRD-UHFFFAOYSA-N
PubChem CID 11572
IUPAC Name 4-methylcyclohexene
SMILES CC1CCC=CC1

Specifications

Color Colorless
Boiling Point 103°C
Quantity 5 g
UN Number 3295
Formula Weight 96.17
Physical Form Liquid
Chemical Name or Material 4-Methyl-1-cyclohexene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.