Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-1-naphthaleneboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. M24575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2457-5G 5 g
M24571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2457-5G Supplier TCI America Supplier No. M24575G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 103986-53-4
Molecular Formula C11H11BO2
Molecular Weight (g/mol) 186.02
MDL Number MFCD01632204
InChI Key JHVQEUGNYSVSDH-UHFFFAOYSA-N
PubChem CID 2773511
IUPAC Name (4-methylnaphthalen-1-yl)boronic acid
SMILES CC1=CC=C(B(O)O)C2=CC=CC=C12

Specifications

Melting Point 207°C
Color Yellow
Quantity 5 g
Synonym 4-methyl-1-naphthaleneboronic acid, 4-methyl-1-naphthalene boronic acid, 4-methylnaphthalene-1-boronic acid, 1-methylnaphthalene-4-boronic acid, 4-methylnaphthalen-1-yl boronic acid
Formula Weight 186.02
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.