Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-1-pentene, 97%

Catalog No. ac127420250
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC127420250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC127420250 Supplier Thermo Scientific Chemicals Supplier No. 127420250
Only null left
Add to Cart

CAS: 691-37-2 | C6H12 | 84.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 691-37-2
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00008949
InChI Key WSSSPWUEQFSQQG-UHFFFAOYSA-N
Synonym 4-methyl-1-pentene, 1-pentene, 4-methyl, poly 4-methyl-1-pentene, unii-x10hrj2y7w, x10hrj2y7w, poly 4-methylpentene, isobutylethene, isohexene, 4-methylpentene-1, 1-pentene, 4-methyl-, homopolymer
PubChem CID 12724
IUPAC Name 4-methylpent-1-ene
SMILES CC(C)CC=C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -155°C
Color Colorless
Density 0.6600g/mL
Boiling Point 53°C to 54°C
Flash Point -31°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.3810 to 1.383
Linear Formula (CH3)2CHCH2CH=CH2
Quantity 25 mL
Specific Gravity 0.66
Formula Weight 84.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Methyl-1-pentene, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 211-720-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.