Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 4-Methyl-1-piperazinecarbonyl chloride hydrochloride, 97%

Catalog No. AAH6675914
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6675914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6675914 Supplier Thermo Scientific™ Supplier No. H6675914
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Chemical Identifiers

CAS 55112-42-0
Molecular Formula C6H12Cl2N2O
Molecular Weight (g/mol) 199.08
MDL Number MFCD01074875
InChI Key WICNYNXYKZNNSN-UHFFFAOYSA-N
IUPAC Name 4-methylpiperazine-1-carbonyl chloride hydrochloride
SMILES Cl.CN1CCN(CC1)C(Cl)=O

Specifications

Melting Point 225-228°
Quantity 25 g
UN Number UN3261
Formula Weight 199.08
Percent Purity 97%
Physical Form Solid
ShelfLife 5 Years
EINECSNumber 259-482-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.