Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-1H-imidazole-5-carbaldehyde, 97%, Thermo Scientific™

Catalog No. SB01366EE
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB01366EE 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB01366EE Supplier Thermo Scientific Chemicals Supplier No. SB01366EE
Add to Cart

Chemical Identifiers

CAS 68282-53-1
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD00173728
InChI Key KMWCSNCNHSEXIF-UHFFFAOYSA-N
Synonym 4-methyl-1h-imidazole-5-carbaldehyde, 5-methylimidazole-4-carboxaldehyde, 4-methylimidazole-5-carboxaldehyde, 4-methyl-5-imidazolecarboxaldehyde, 5-methyl-3h-imidazole-4-carbaldehyde, 4-methylimidazole-5-carbaldehyde, 1h-imidazole-4-carboxaldehyde, 5-methyl, 1h-imidazole-5-carboxaldehyde, 4-methyl, pubchem23016, acmc-1b6zs
PubChem CID 2795887
IUPAC Name 5-methyl-1H-imidazole-4-carbaldehyde
SMILES CC1=C(C=O)N=CN1

Specifications

Assay Percent Range 97%
Quantity 50 g
Formula Weight 110.12
Percent Purity 97%
Chemical Name or Material 4-Methyl-1H-imidazole-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.