Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2-(2-pyrazinyl)-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC19801DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC19801DA 1 g
CC19801CB 250 mg
CC19801DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC19801DA Supplier Thermo Scientific Chemicals Supplier No. CC19801DA
Add to Cart

Chemical Identifiers

CAS 216959-92-1
Molecular Formula C9H7N3O2S
Molecular Weight (g/mol) 221.23
MDL Number MFCD00111662
InChI Key XDZXQDHRHGIPRN-UHFFFAOYSA-N
Synonym 4-methyl-2-2-pyrazinyl-1,3-thiazole-5-carboxylic acid, 4-methyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid, 2-5-carboxy-4-methyl-1,3-thiazol-2-yl pyrazine, 4-methyl-2-2-pyrazinyl thiazole-5-carboxylic acid, 4-methyl-2-2-pyrazinyl-thiazole-5-carboxylic acid, 4-methyl-2-pyrazin-2-yl thiazole-5-carboxylic acid, 5-carboxy-4-methyl-2-pyrazin-2-yl-1,3-thiazole, 5-thiazolecarboxylicacid, 4-methyl-2-2-pyrazinyl, 5-thiazolecarboxylic acid, 4-methyl-2-2-pyrazinyl
PubChem CID 2776505
IUPAC Name 4-methyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC(=N1)C1=NC=CN=C1)C(O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 221.24
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-(2-pyrazinyl)-1, 3-thiazole-5-carboxylic acid
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.