Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Methyl-2,3-dihydrocyclopenta[b]indol-1(4H)-one | 50776-26-6 | 99.9% | C12H11NO | 5 G
SDP

Catalog No. 5000433587
Encompass_Preferred

4-Methyl-2,3-dihydrocyclopenta[b]indol-1(4H)-one is a chemical compound used as a drug intermediate for the synthesis of various active compounds. This product has a high purity of 99.9% and appears as a light yellow to yellow solid.

  • Targeted as a drug intermediate
  • Chemical formula: C12H11NO
  • Molecular weight: 185.23
  • Purity (LCMS): 99.9%
  • Appearance: Light yellow to yellow solid
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-33587 Supplier Medchemexpress LLC Supplier No. HY753575G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Methyl-2,3-dihydrocyclopenta[b]indol-1(4H)-one is a chemical compound used as a drug intermediate for the synthesis of various active compounds. This product has a high purity of 99.9% and appears as a light yellow to yellow solid.

  • Targeted as a drug intermediate
  • Chemical formula: C12H11NO
  • Molecular weight: 185.23
  • Purity (LCMS): 99.9%
  • Appearance: Light yellow to yellow solid
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.