Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-methyl-2-(4-methylpiperazinyl)pyrimido[4,5-b]benzothiazine | 928853-86-5 | 98.5% | 313.42 | C16H19N5S | 100 MG
SDP

Catalog No. 5000307833
Encompass_Preferred

4-MMPB (CAS 928853-86-5) is a small-molecule research compound that selectively inhibits 15-lipoxygenase and has been used in studies of cancer biology. It is provided as a high-purity research reagent with characterized activity and recommended storage conditions for long-term stability.

  • Selective 15-lipoxygenase inhibitor with reported IC50 ≈ 18 μM.
  • High purity suitable for research (≈98.5%).
  • Available in small pack sizes for laboratory use, including solid and solution formats.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Molecular formula C16H19N5S and molecular weight 313.42, suitable for analytical characterization.

Catalog No. 50-003-07833 Supplier Medchemexpress LLC Supplier No. HY118480100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-MMPB (CAS 928853-86-5) is a small-molecule research compound that selectively inhibits 15-lipoxygenase and has been used in studies of cancer biology. It is provided as a high-purity research reagent with characterized activity and recommended storage conditions for long-term stability.

  • Selective 15-lipoxygenase inhibitor with reported IC50 ≈ 18 μM.
  • High purity suitable for research (≈98.5%).
  • Available in small pack sizes for laboratory use, including solid and solution formats.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Molecular formula C16H19N5S and molecular weight 313.42, suitable for analytical characterization.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.