Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2-nitroaniline 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
89-62-3
C7H8N2O2
152.15
MFCD00007907
DLURHXYXQYMPLT-UHFFFAOYSA-N
2-nitro-p-toluidine,4-amino-3-nitrotoluene,3-nitro-4-toluidine,fast red gl,3-nitro-4-aminotoluene,fast red gl base,benzenamine, 4-methyl-2-nitro,2-nitro-4-methylaniline,4-methyl-6-nitroaniline,mnpt
6978
CHEBI:66920
4-methyl-2-nitroaniline
CC1=CC(=C(C=C1)N)[N+](=O)[O-]
89-62-3
C7H8N2O2
152.15
MFCD00007907
DLURHXYXQYMPLT-UHFFFAOYSA-N
2-nitro-p-toluidine,4-amino-3-nitrotoluene,3-nitro-4-toluidine,fast red gl,3-nitro-4-aminotoluene,fast red gl base,benzenamine, 4-methyl-2-nitro,2-nitro-4-methylaniline,4-methyl-6-nitroaniline,mnpt
6978
CHEBI:66920
4-methyl-2-nitroaniline
CC1=CC(=C(C=C1)N)[N+](=O)[O-]
Boiling Point 169°C (21.0mmHg)
Melting Point 112°C to 115°C
Flash Point 157°C
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Color Brown-Orange to Orange
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3C6H3(NO2)NH2
Beilstein 12, 1000
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and diethyl ether
Formula Weight 152.15
Physical Form Fine Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 4-Methyl-2-nitroaniline, 99%
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
Toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-924-9

RTECSNumber : XU8227250

TSCA : TSCA