Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2-nitroaniline 98.0+%, TCI America™
SDP

Catalog No. N011425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N011425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N011425G Supplier TCI America Supplier No. N011425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 89-62-3
Melting Point 116°C
Molecular Formula C7H8N2O2
Quantity 25 g
MDL Number MFCD00007907
UN Number 2660
Synonym 2-nitro-p-toluidine, 4-amino-3-nitrotoluene, 3-nitro-4-toluidine, fast red gl, 3-nitro-4-aminotoluene, fast red gl base, benzenamine, 4-methyl-2-nitro, 2-nitro-4-methylaniline, 4-methyl-6-nitroaniline, mnpt
InChI Key DLURHXYXQYMPLT-UHFFFAOYSA-N
SMILES CC1=CC=C(N)C(=C1)[N+]([O-])=O
IUPAC Name 4-methyl-2-nitroaniline
Molecular Weight (g/mol) 152.15
PubChem CID 6978
ChEBI CHEBI:66920
Formula Weight 152.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-2-nitroaniline
Show More Show Less
EINECSNumber (3)-0401&(5)-2302
RTECSNumber XU8227250
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.