Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2-phenyl-1,3-thiazole-5-carbaldehyde, Thermo Scientific™

Catalog No. CC29204DA
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC29204DA 1 g
CC29204CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC29204DA Supplier Thermo Scientific Chemicals Supplier No. CC29204DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 55327-23-6
Molecular Formula C11H9NOS
Molecular Weight (g/mol) 203.26
MDL Number MFCD04974047
InChI Key QRPSQJOXOZJEHJ-UHFFFAOYSA-N
Synonym 4-methyl-2-phenylthiazole-5-carbaldehyde, 4-methyl-2-phenyl-thiazole-5-carbaldehyde, 2-phenyl-4-methylthiazole-5-carbaldehyde, 5-thiazolecarboxaldehyde, 4-methyl-2-phenyl
PubChem CID 2795491
IUPAC Name 4-methyl-2-phenyl-1,3-thiazole-5-carbaldehyde
SMILES CC1=C(SC(=N1)C1=CC=CC=C1)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 203.26
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-phenyl-1, 3-thiazole-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.