Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Methyl-2-(pyridin-3-yl)thiazole-5-carboxylic acid | 39091-01-5 | 96.8% | 220.25 | 1 G
SDP

Catalog No. 5000453695
Encompass_Preferred

2-(3-Pyridyl)-4-methyl-thiazole-5-carboxylic acid is a synthetic intermediate useful for pharmaceutical synthesis.

  • Synthetic intermediate for pharmaceutical applications
  • Appearance: White to off-white solid
  • Purity (HPLC): 96.81%
  • Molecular Formula: C10H8N2O2S
  • Molecular Weight: 220.25

Catalog No. 50-004-53695 Supplier Medchemexpress LLC Supplier No. HYW0124281G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(3-Pyridyl)-4-methyl-thiazole-5-carboxylic acid is a synthetic intermediate useful for pharmaceutical synthesis.

  • Synthetic intermediate for pharmaceutical applications
  • Appearance: White to off-white solid
  • Purity (HPLC): 96.81%
  • Molecular Formula: C10H8N2O2S
  • Molecular Weight: 220.25

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.