Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Methyl-2-(pyridin-3-yl)thiazole-5-carboxylic acid | 39091-01-5 | 96.8% | 220.25 | 250MG
SDP

Catalog No. 5000451361
Encompass_Preferred

2-(3-Pyridyl)-4-methyl-thiazole-5-carboxylic acid is a synthetic intermediate useful for pharmaceutical synthesis.

  • Target: Drug intermediate
  • Pathway: Others
  • Storage: Please store the product under the recommended conditions in the Certificate of Analysis.

Catalog No. 50-004-51361 Supplier Medchemexpress LLC Supplier No. 250MGHYW012428
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(3-Pyridyl)-4-methyl-thiazole-5-carboxylic acid is a synthetic intermediate useful for pharmaceutical synthesis.

  • Target: Drug intermediate
  • Pathway: Others
  • Storage: Please store the product under the recommended conditions in the Certificate of Analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.