Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-3,5-dinitrobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. D329125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D329125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D329125G Supplier TCI America Supplier No. D329125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16533-71-4
Molecular Formula C8H6N2O6
Molecular Weight (g/mol) 226.14
MDL Number MFCD00007175
InChI Key LZWWZQXBKVZKIP-UHFFFAOYSA-N
Synonym 4-methyl-3,5-dinitrobenzoic acid, 3,5-dinitro-4-methylbenzoic acid, 3,5-dinitro-p-toluic acid, 3,5-dinitro-4-methyl benzoic acid, benzoic acid, 4-methyl-3,5-dinitro, p-toluic acid, 3,5-dinitro, acmc-209dsg, p-toluic acid,5-dinitro, ksc496a8j, 3,5-dinitro-4-methylbenzoicacid
PubChem CID 85477
IUPAC Name 4-methyl-3,5-dinitrobenzoic acid
SMILES CC1=C(C=C(C=C1[N+]([O-])=O)C(O)=O)[N+]([O-])=O

Specifications

Melting Point 160°C
Color White-Yellow
Quantity 25 g
Formula Weight 226.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-3,5-dinitrobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.