Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-methyl-3-nitrobenzoic acid | 96-98-0 | MFCD00007174 | 100.0% | 181.15 g·mol-1 | C8H7NO4 | 1 KG
SDP

Catalog No. 5000301992
Encompass_Preferred

4-Methyl-3-nitrobenzoic acid is an organic reagent used as a synthetic building block and biochemical reagent in life-science and chemical research. It is supplied as a solid with documented analytical data and safety information to support laboratory use.

  • High purity validated by HPLC (99.96% reported on COA).
  • Molecular formula C8H7NO4 and molecular weight 181.15 g·mol-1.
  • Available in multiple pack sizes, including 1 kg, for scale flexibility.
  • Certificate of analysis and safety data sheet available for traceability.
  • Recommended storage conditions provided to preserve stability.
  • Suitable as an intermediate in organic synthesis and for research applications.

Catalog No. 50-003-01992 Supplier Medchemexpress LLC Supplier No. HYW0212931KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Methyl-3-nitrobenzoic acid is an organic reagent used as a synthetic building block and biochemical reagent in life-science and chemical research. It is supplied as a solid with documented analytical data and safety information to support laboratory use.

  • High purity validated by HPLC (99.96% reported on COA).
  • Molecular formula C8H7NO4 and molecular weight 181.15 g·mol-1.
  • Available in multiple pack sizes, including 1 kg, for scale flexibility.
  • Certificate of analysis and safety data sheet available for traceability.
  • Recommended storage conditions provided to preserve stability.
  • Suitable as an intermediate in organic synthesis and for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.