Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-3-nitropyridine 97.0+%, TCI America™
SDP

Catalog No. M23535G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23535G 5 g
M23531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23535G Supplier TCI America Supplier No. M23535G

Chemical Identifiers

CAS 5832-44-0
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00051829
InChI Key JLNRJMGYBKMDGI-UHFFFAOYSA-N
Synonym 3-nitro-4-picoline, 4-methyl-3-nitro-pyridine, 3-nitro-4-methylpyridine, pyridine, 4-methyl-3-nitro, zlchem 957, pubchem6644, 4-methyl-3-nitro pyridine, acmc-1b13v, ksc490s2l, tpc-py011
PubChem CID 521828
IUPAC Name 4-methyl-3-nitropyridine
SMILES CC1=C(C=NC=C1)[N+](=O)[O-]

Specifications

Color Yellow
Boiling Point 110°C
Quantity 5 g
Formula Weight 138.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Methyl-3-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.