Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-3-penten-2-one, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019505 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9505 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9505 Supplier MilliporeSigma Supelco Supplier No. 497221ML
Add to Cart
Add to Cart

4-Methyl-3-penten-2-one belongs to the volatile organic compounds (VOCs) class of chemicals.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 141-79-7
Density 0.858 g/mL (at 25°C)
Flash Point 31°C - Closed Cup
Physical Form Neat
Linear Formula (CH3)2 C=CHCOCH3
MDL Number MFCD00008900
Melting Point -41.5°C
Boiling Point 130°C
Synonym Mesityl oxide; 4-Methyl-3-penten-2-one
Type Analytical Standard
Quantity 1 mL
Formula Weight 98.14
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.