Learn More
eMolecules​ 1227833-81-9 | 4-Methyl-3-pyridylzinc bromide, 0.50 M in THF | Synthonix237.410 | C6H6BrNZn | 0.000 | [Br-].Cc1ccncc1[Zn+] | 25ml | 627353705
Catalog No. 502695274 Shop All eMolecules Inc. Products
Encompass_Preferred
4-Methyl-3-pyridylzinc bromide, 0.50 M in THF | Synthonix | 1227833-81-9237.410 | C6H6BrNZn | 0.000 | [Br-].Cc1ccncc1[Zn+] | 25ml | 627353705
Catalog No. 50-269-5274 Supplier eMolecules​ Supplier No. 627353705
4-Methyl-3-pyridylzinc bromide, 0.50 M in THF | Synthonix | 1227833-81-9237.410 | C6H6BrNZn | 0.000 | [Br-].Cc1ccncc1[Zn+] | 25ml | 627353705