Learn More
Medchemexpress LLC 4-methyl-5-[2-(4-morpholin-4-ylanilino)pyrimidin-4-yl]-1,3-thiazol-2-amine | 693228-63-6 | MFCD14155805 | 99.1% | 368.46 | C18H20N6OS | 100 MG
SDP
Catalog No. 5000240167
Encompass_Preferred
CYC-116 is a small-molecule research inhibitor of Aurora kinases A and B with low-nanomolar potency. It is used in biochemical and cellular studies to probe cell cycle regulation and antitumor activity, and exhibits additional activity against VEGFR2, Src, Lck, and FLT3.
  • Potent inhibitor of Aurora A and B (Ki ≈ 8 and 9 nM).
  • Additional kinase activity against VEGFR2, Src, Lck, and FLT3.
  • Defined molecular formula and weight: C18H20N6OS, 368.46 g/mol.
  • High reported purity (>99% by HPLC) suitable for research applications.
  • Supplied in 100 mg research quantities for laboratory use.
Catalog No. 50-002-40167 Supplier Medchemexpress LLC Supplier No. HY10558100MG
CYC-116 is a small-molecule research inhibitor of Aurora kinases A and B with low-nanomolar potency. It is used in biochemical and cellular studies to probe cell cycle regulation and antitumor activity, and exhibits additional activity against VEGFR2, Src, Lck, and FLT3.
  • Potent inhibitor of Aurora A and B (Ki ≈ 8 and 9 nM).
  • Additional kinase activity against VEGFR2, Src, Lck, and FLT3.
  • Defined molecular formula and weight: C18H20N6OS, 368.46 g/mol.
  • High reported purity (>99% by HPLC) suitable for research applications.
  • Supplied in 100 mg research quantities for laboratory use.