Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-methyl-5-[2-(4-morpholin-4-ylanilino)pyrimidin-4-yl]-1,3-thiazol-2-amine | 693228-63-6 | MFCD14155805 | 99.1% | 368.46 g/mol | C18H20N6OS | 1 ML
SDP

Catalog No. 5000240176
Encompass_Preferred

CYC-116 is a small-molecule inhibitor of Aurora A and Aurora B kinases for laboratory research. It exhibits low-nanomolar affinity (Ki ≈ 8 nM for Aurora A; 9 nM for Aurora B) and is supplied as a high-purity solid or as a 10 mM solution in DMSO for in vitro studies. Intended for research use only.

  • Potent Aurora A and B inhibition with Kis of 8 nM and 9 nM.
  • High purity (≈99.1%) suitable for biochemical assays.
  • Available as solid and 10 mM 1 mL solution in DMSO.
  • Molecular formula C18H20N6OS; molecular weight 368.46 g/mol.
  • Storage: powder -20°C (up to 3 years); solution -80°C (recommended).
  • Intended for in vitro research use only.

Catalog No. 50-002-40176 Supplier Medchemexpress LLC Supplier No. HY1055810MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CYC-116 is a small-molecule inhibitor of Aurora A and Aurora B kinases for laboratory research. It exhibits low-nanomolar affinity (Ki ≈ 8 nM for Aurora A; 9 nM for Aurora B) and is supplied as a high-purity solid or as a 10 mM solution in DMSO for in vitro studies. Intended for research use only.

  • Potent Aurora A and B inhibition with Kis of 8 nM and 9 nM.
  • High purity (≈99.1%) suitable for biochemical assays.
  • Available as solid and 10 mM 1 mL solution in DMSO.
  • Molecular formula C18H20N6OS; molecular weight 368.46 g/mol.
  • Storage: powder -20°C (up to 3 years); solution -80°C (recommended).
  • Intended for in vitro research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.