Learn More
Medchemexpress LLC 4-methyl-5-[2-(4-morpholin-4-ylanilino)pyrimidin-4-yl]-1,3-thiazol-2-amine | 693228-63-6 | MFCD14155805 | 99.1% | 368.46 g·mol⁻¹ | C18H20N6OS | 25 MG
SDP
Catalog No. 5000241055
Encompass_Preferred
CYC-116 is a small-molecule inhibitor of Aurora A and Aurora B kinases with low-nanomolar inhibitory potency. It is used as a research reagent to investigate mitotic signaling and cell-cycle regulation and is supplied as a purified solid for in vitro studies.
  • Potent dual Aurora A and B inhibitor with Ki ≈8-9 nM.
  • Suitable for studying mitotic signaling and cell-cycle regulation in vitro.
  • High purity (>99.0%) for reliable experimental results.
  • Chemical formula C18H20N6OS; molecular weight 368.46 g·mol⁻¹.
  • Available in multiple small-quantity pack sizes for laboratory use.
Catalog No. 50-002-41055 Supplier Medchemexpress LLC Supplier No. HY1055825MG
CYC-116 is a small-molecule inhibitor of Aurora A and Aurora B kinases with low-nanomolar inhibitory potency. It is used as a research reagent to investigate mitotic signaling and cell-cycle regulation and is supplied as a purified solid for in vitro studies.
  • Potent dual Aurora A and B inhibitor with Ki ≈8-9 nM.
  • Suitable for studying mitotic signaling and cell-cycle regulation in vitro.
  • High purity (>99.0%) for reliable experimental results.
  • Chemical formula C18H20N6OS; molecular weight 368.46 g·mol⁻¹.
  • Available in multiple small-quantity pack sizes for laboratory use.