Learn More
Medchemexpress LLC 4-methyl-5-thiazole ethanol | 137-00-8 | MFCD00005339 | 99.5% | 143.21 | C6H9NOS | 25 G
SDP
Catalog No. 5000303453
Encompass_Preferred
4-Methyl-5-thiazoleethanol (CAS 137-00-8) is a sulfur-containing thiazole alcohol used as a flavor compound with a beef and nut aroma and as a reagent and analytical standard. It is supplied as a high-purity liquid (≈99.5%), molecular formula C6H9NOS, molecular weight 143.21 g/mol, and is available in laboratory and bulk pack sizes with COA and SDS documentation available.
  • High purity suitable for analytical and research use.
  • Characteristic beef and nut aroma for flavor applications.
  • Supplied as a liquid with documented physical and chemical data.
  • COA and SDS available for quality and safety verification.
  • Available in small and bulk pack sizes for flexibility.
Catalog No. 50-003-03453 Supplier Medchemexpress LLC Supplier No. 25GHYW015695
4-Methyl-5-thiazoleethanol (CAS 137-00-8) is a sulfur-containing thiazole alcohol used as a flavor compound with a beef and nut aroma and as a reagent and analytical standard. It is supplied as a high-purity liquid (≈99.5%), molecular formula C6H9NOS, molecular weight 143.21 g/mol, and is available in laboratory and bulk pack sizes with COA and SDS documentation available.
  • High purity suitable for analytical and research use.
  • Characteristic beef and nut aroma for flavor applications.
  • Supplied as a liquid with documented physical and chemical data.
  • COA and SDS available for quality and safety verification.
  • Available in small and bulk pack sizes for flexibility.