Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-D-phenylalanine, 98%, Thermo Scientific™

Catalog No. AAH2751803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2751803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2751803 Supplier Thermo Scientific Chemicals Supplier No. H2751803
  • $59.40
    $50.65 / Each of 1

    Save $8.75

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49759-61-7
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00152137
InChI Key DQLHSFUMICQIMB-UHFFFAOYNA-N
Synonym 4-methyl-d-phenylalanine, h-d-phe 4-me-oh, r-2-amino-3-p-tolyl propanoic acid, d-4-methylphenylalanine, d-4-me-phe-oh, 2r-2-amino-3-4-methylphenyl propanoic acid, d-phenylalanine, 4-methyl, h-p-me-phe-oh, 4-methylphenyl-d-alanine, d-4-methylphe
PubChem CID 7015862
IUPAC Name (2R)-2-amino-3-(4-methylphenyl)propanoic acid
SMILES CC1=CC=C(CC(N)C(O)=O)C=C1

Specifications

Quantity 1 g
Formula Weight 179.22
Percent Purity 98%
Chemical Name or Material 4-Methyl-D-phenylalanine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.