Learn More
Cayman Chemical 4-methyl Erlotinib, Purity: ≥95%, CAS: 1346601-52-2, 10 mg
SDP
Catalog No. 502892948
Encompass_Preferred
A methylated derivative of erlotinib; has been used as an internal standard for the quantification of erlotinib and erlotinib metabolites in human plasma
Catalog No. 50-289-2948 Supplier Cayman Chemical Supplier No. 900151010 MG
A methylated derivative of erlotinib; has been used as an internal standard for the quantification of erlotinib and erlotinib metabolites in human plasma